M13 Skeletal Muscle Myosin Light Chain Kinase Peptide
Supplier: Anaspec Inc
AS-60184-1
AS-60184-5
103002-830EA
380
USD
103002-830
103002-832
M13 Skeletal Muscle Myosin Light Chain Kinase Peptide
Proteins and Peptides
M-13 is a peptide that represents CAM-binding domain of Calmodulin (CaM) target proteins. CaM is an ubiquitous Ca2+ binding protein.
Sequence:KRRWKKNFIAVSAANRFKKISSSGAL
MW:2964.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...