Human Recombinant MMP-1 (from E. coli)
Supplier: Anaspec Inc
AS-55575-1 AS-55575-10
103001-682EA 182.69 USD
103001-682 103001-684
Human Recombinant MMP-1 (from E. coli)
Proteins and Peptides
Matrix metalloproteinases (MMP’s) belong to a family of secreted or membrane-associated zinc endopeptidases capable of digesting extracellular matrix components. MMP-1 (collagenase-1) is involved in tumor development and metastasis and rheumatoid arthritis. It is proposed as a therapeutic target for these diseases. MMP-1 digests a broad range of substrates, including α-1 antitrypsin, myelin basic protein, collagen I, II, III, VII, VIII, casein, gelatin, and others
The sequence (Accession # NP_002412) corresponding to the catalytic domain (aa 106-261) of Human MMP-1 was expressed in E. coli. The recombinant human MMP-1 was purified from bacterial lysate and refolded using proprietary technique. The molecular weight of the recombinant Human MMP-1 Catalytic Domain is ~17.5 kDa.
17.5 kDa.