Human Recombinant Tau (from E. coli)
Supplier: Anaspec Inc
AS-55556-50
103001-650EA 403.38 USD
103001-650
Human Recombinant Tau (from E. coli)
Proteins and Peptides
The sequence (Accession # AAC04279.1) corresponding to the full length human Tau-441 (2N4R isoform) protein along with GST tag was expressed in E. coli. The recombinant Tau-441 protein was purified from bacterial lysate using proprietary method. GST tag was cleaved and removed using Glutathione agarose.
Applications: In vitro phosphorylation and acetylation, Western immunoblotting and ELISA standard.
Microtubule associated protein (Tau) is found predominantly in the central neural system and its major function is to promote assembly and to stabilize neuronal microtubules. Six isoforms of Tau were identified in humans that are differentiated by the exclusion or inclusion of exons 2, 3, and 10. Tau-441 is the longest of Tau isoforms, consisting of 441 amino acids with molecular mass of 45.8 kDa. Under physiological conditions Tau can undergo abnormal phosphorylation, truncation, or other modifications that result in the protein detachment from microtubules. These modified Tau molecules can self-associate and form different types of aggregates including neurofibrillary tangles (NFTs) found in brains of patients with neurodegenerative diseases such as Alzheimer’s disease.