Influenza CEF Influenza
Supplier: Anaspec Inc
AS-28334
103000-722EA
187.08
USD
103000-722
Influenza CEF Influenza
Proteins and Peptides
HLA-A*03 restricted epitope from influenza virus nucleoprotein (265-274).
Sequence: ILRGSVAHK
MW: 980.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...