Human Beta-Amyloid (1-40)

Supplier: Anaspec Inc

AS-24235- AS-24236 AS-24236-5
102999-790EA 247 USD
102999-790 102999-792 102999-794
Human Beta-Amyloid (1-40)
Proteins and Peptides

Aß (1-40) together with Aß (1-42) are two major C-terminal variants of the Aß protein constituting the majority of Aßs. These undergo post-secretory aggregation and deposition in the Alzheimer’s disease brain.
Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV
Molecular Weight: 4329.9 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR