Human;Rat Biotin-Corticotropin Releasing Factor, Biotin-CRF,Biotin
Supplier: Anaspec Inc
AS-23976
102999-772EA
571.46
USD
102999-772
Human;Rat Biotin-Corticotropin Releasing Factor, Biotin-CRF,Biotin
Proteins and Peptides
The peptide, corticotropin releasing factor (CRF) was first isolated in the mammalian brain that regulates the hypothalamic-pituitary-adrenocortical axis. It also plays a key role in modulating the endocrine, autonomic, and behavioral responses to stress and cardiovascular, gastrointestinal, and immune activities.
Sequence: Biotin-SEEPPISLDLTFHLLREVLEMARAEQLAQQAHSNRKLMEII-NH2
MW: 4984.8 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...