Beta-Secretase Inhibitor 1
Supplier: Anaspec Inc
AS-23958
102999-752EA
339.08
USD
102999-752
Beta-Secretase Inhibitor 1
Proteins and Peptides
ß-Secretase (BACE1) is a key enzyme involved in the production of Aß peptides found in extracellular amyloid plaques of Alzheimer’s disease (AD). The enzyme has been implicated as an excellent target for anti-amyloid therapy of AD. This statine-based substrate analog, beta-secretase inhibitor P10–P4’ statV, is used in the inhibition of beta-secretase activity from homogenized wild-type mouse cortices and from BACE-purified from human brain.
Sequence:KTEEISEVN-Sta-VAEF (Sta = statine)
MW:1651.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...