Human Beta-Amyloid (11-22)
Supplier: Anaspec Inc
AS-23956
102999-750EA
194.38
USD
102999-750
Human Beta-Amyloid (11-22)
Proteins and Peptides
This is amino acids 11 to 22 fragment of the beta-amyloid peptide.
Sequence: EVHHQKLVFFAE
Molecular Weight: 1483.7 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...