Human Beta-Amyloid (1-42), Biotin
Supplier: Anaspec Inc
This is biotinylated Beta-Amyloid (1-42) peptide, which has been used in studies to examine aggregation/polymerization effects in the presence of drug candidates.
Sequence: Biotin-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
MW: 4740.7 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Citations:
Mandler, M. et al. (2015). Tailoring the antibody response to aggregated Aß using novel Alzheimer-Vaccines PLoS One doi: 10.1371/journal.pone.0115237.
Abraham, JD. et al. (2013). Cerebrospinal Aβ11-x and 17-x levels as indicators of mild cognitive impairment and patients’ stratification in Alzheimer’s disease. Transl Psychiatry 3, e281. doi: 10.1038/tp.2013.58.
Arriagada, C. et al. (2010). Apoptosis is directly related to intracellular amyloid accumulation in a cell line derived from the cerebral cortex of a trisomy 16 mouse, an animal model of Down syndrome. Neuro Lett 470, 81. doi:10.1016/j.neulet.2009.12.062.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...