Human [Gla17,21,24]-Osteocalcin (1-49) Peptide

Supplier: Anaspec Inc

AS-22829 AS-22830 AS-22831
102999-354EA 251.38 USD
102999-354 102999-356 102999-358
Human [Gla17,21,24]-Osteocalcin (1-49) Peptide
Proteins and Peptides

Osteocalcin (OC) is a 49 amino acid peptide found exclusively in bone tissue and is highly conserved among species. It is a vitamin K- and D-dependent protein produced by osteoblasts, osteocytes and odontoblasts. It is deposited in extracellular bone matrix and is found in the serum. Serum osteocalcin, hydrolysed in the kidney and liver, is considered a specific marker of osteoblast activity and bone formation rate. It may be involved in regulation of osteoblast function, regulation of bone turnover and/or mineralization.
Sequence: YLYQWLGAPVPYPDPL-Gla-PRR-Gla-VC-Gla-LNPDCDELADHIGFQEAYRRFYGPV (Gla=γ-Carboxyglutamic Acid; Disulfide bridge: 23-29)
MW: 5929.5 Da
% Peak area by HPLC: 95
Storage condition: -20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR