VSV VSV-G Peptide
Supplier: Anaspec Inc
AS-21152
102998-830EA
108.15
USD
102998-830
VSV VSV-G Peptide
Proteins and Peptides
A vesicular stomatitis virus G (VSV-G) protein fragment.
Sequence:YTDIEMNRLGK
MW:1339.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...