Human Calcitonin Gene Related Peptide
Supplier: Anaspec Inc
AS-20682
AS-20681
102996-084EA
521.77
USD
102996-084
102998-458
Human Calcitonin Gene Related Peptide
Proteins and Peptides
CGRP is a 37-amino acid neuropeptide produced by tissue specific processing of the calcitonin gene and is the major product in neural tissues.
Sequence:ACDTATCVTHRLAGLLSRSGGVVKNNFVPTNVGSKAF-NH2 (Disulfide bridge: 2-7)
MW:3789.4 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...