Human Calcitonin Peptide
Supplier: Anaspec Inc
AS-20673
AS-20674
102998-448EA
268.92
USD
102998-448
102998-450
Human Calcitonin Peptide
Proteins and Peptides
Human calcitonin reduces blood calcium, opposing the effects of parathyroid hormone (PTH). It stimulates cyclic nucleotide accumulation in human kidney cortex and medulla. Calcitonin maybe involved in osteoporosis and in Paget's disease.
Sequence: CGNLSTCMLGTYTQDFNKFHTFPQTAIGVGAP-NH2 (Disulfide bridge: 1-7)
MW: 3417.9 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...