Human Bradykinin
Supplier: Anaspec Inc
AS-20667
102998-444EA
61.38
USD
102998-444
Human Bradykinin
Proteins and Peptides
This peptide is an inhibitor for Angiotensin I Converting Enzyme (ACE I), derived from Bradykinin. ACE I partially suppresses the renin-angiotensin-aldosterone system (RAAS), which regulates blood pressure and may mediate hypertension. ACE I converts angiotensin I to the biologically active peptide angiotensin II using a zinc- and chloride- dependent mechanism.
Sequence: RPPGFSPFR
MW: 1060.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...