Human ACTH (1-10)
Supplier: Anaspec Inc
AS-20624
102998-434EA
182.69
USD
102998-434
Human ACTH (1-10)
Proteins and Peptides
Centrally administered N-terminal fragments of ACTH (1-10, 4-10, 4-9) display convulsant properties in rabbits.
Sequence: SYSMEHFRWG
MW: 1299.4 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...