Human Recombinant MOG (1-125) (from E. coli)
Supplier: Anaspec Inc
AS-55158-100 AS-55158-1000 AS-55158-500
102998-096EA 374.15 USD
102998-096 102998-098 102998-100
Human Recombinant MOG (1-125) (from E. coli)
Proteins and Peptides
Myelin Oligodendrocyte Glycoprotein (MOG) is a member of the immunoglobulin superfamily and is expressed exclusively in the central nervous system (CNS). Although MOG protein constitutes only 0.01-0.05% of the CNS myelin proteins, it was demonstrated that MOG protein is a crucial autoantigen for multiple sclerosis in humans and experimental autoimmune encephalomyelitis (EAE) in rodents and monkeys
The sequence (Accession # CAQ10087) corresponding to the extracellular domain of human MOG along with a 6x His tag was expressed in E. coli. The recombinant human MOG (H-rMOG) was purified from urea denatured bacterial lysate using immobilized metal affinity chromatography (IMAC). The molecular mass of the recombinant human MOG is 14.2 kDa