AKT/PKB/Rac-Protein Kinase Substrate, AnaSpec
Supplier: Anaspec Inc
AS-29945-1
AS-29945-5
102997-486EA
279.15
USD
102997-486
102997-488
AKT/PKB/Rac-Protein Kinase Substrate, AnaSpec
Proteins and Peptides
The native peptide ARKRERTYSFGHHA (29944-1) is a synthetic substrate for AKT/PKB/Rac-protein kinases. Phophorylation is at the Ser site (Km = 3.9 µM). It also competitively inhibits histone H2B phosphorylation (Ki = 12 µM) by AKT.
Sequence:Biotin-ARKRERTYSFGHHA
MW:1942.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...