Tyrosine Kinase Peptide 1, TMR (Tetramethylrhodamine)
Supplier: Anaspec Inc
AS-29935-1
102997-410EA
663.54
USD
102997-410
Tyrosine Kinase Peptide 1, TMR (Tetramethylrhodamine)
Proteins and Peptides
Peptide sequence KVEKIGEGTYGVVYK is derived from the amino acid residues CDC26-20. It is considered to be a generic substrate for various TPKs.
Sequence:5-TMR-KVEKIGEGTYGVVYK
MW:2082.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...