Human [Pyr3]-beta-Amyloid (3-42)
Supplier: Anaspec Inc
AS-29907-01
AS-29907-1-
102997-272EA
201.69
USD
102997-272
102997-274
Human [Pyr3]-beta-Amyloid (3-42)
Proteins and Peptides
This is one of the predominant amyloid peptide structures deposited in human brain of Alzheimer’s disease and Down’s syndrome patients.Sequence: Pyr-FRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
Molecular Weight: 4309.9 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...