Human [Pyr3]-beta-Amyloid (3-40)
Supplier: Anaspec Inc
AS-29906-01 AS-29906-1-
102997-268EA 157.85 USD
102997-268 102997-270
Human [Pyr3]-beta-Amyloid (3-40)
Proteins and Peptides
This peptide triggers the formation of insoluble Aß peptide deposits, inhibits secretion of amyloid protein precursor (APP) and increases cell-associated APP. The fibrils in the plaques formed in the brains of patients with Alzheimer’s disease contain significant amounts of truncated Aß proteins such as Aß(3-40) and Aß (11- 40). These two fragments contain an N-terminal pyroglutamic acid. The g-carboxy group of glutamic acid forms a cyclic bond with the free N-terminal amino group to form pyroglutamate.
  • Peptide Content: ≥ 60%