Human Beta-Amyloid (2-40)
Supplier: Anaspec Inc
AS-29905-1
102997-266EA
726.38
USD
102997-266
Human Beta-Amyloid (2-40)
Proteins and Peptides
This peptide is beta-amyloid (1-40) N-terminally truncated. It was shown that supplementing the media with N-terminally truncated Abeta (2-40) and (2-42) induce the phagocytosis of polystyrene particles by primary human monocytes. N-terminally truncated Aβ(x–42) induced the phagocytosis of PSPs significantly more effectively than did Aβ(x–40).
Sequence: AEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV
Molecular Weight: 4214.8 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...