EBV (Epstein-Barr virus) CEF EBV
Supplier: Anaspec Inc
AS-28320
102997-158EA
187.08
USD
102997-158
EBV (Epstein-Barr virus) CEF EBV
Proteins and Peptides
HLA A2.1-restricted epitope from Epstein-Barr Virus lytic protein BMLF1 (280-288).
Sequence: GLCTLVAML
MW: 920.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...