S6 Kinase Substrate (229-239),Biotin
Supplier: Anaspec Inc
AS-27193
102996-888EA
251.38
USD
102996-888
S6 Kinase Substrate (229-239),Biotin
Proteins and Peptides
This is a biotinylated synthetic peptide substrate for S6 kinase.
Sequence:Biotin-AKRRRLSSLRA-NH2
MW:1538.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...