Human;Rat [Leu31, Pro34]-Neuropeptide Y
Supplier: Anaspec Inc
AS-24366
102996-554EA
339.08
USD
102996-554
Human;Rat [Leu31, Pro34]-Neuropeptide Y
Proteins and Peptides
Like neuropeptide Y (NPY) and other peptides of the family, this peptide adopts a folded hairpin structure with the terminal segments in close proximity. An intracerebroventricular injection of NPY or [Leu31, Pro34]-NPY (non-Y2 receptor agonist) given during middle cerebral artery occlusion increases the infarct volume and nitric oxide (NO) overproduction in the rat brain.
Sequence:YPSKPDNPGEDAPAEDMARYYSALRHYINLLTRPRY-NH2
MW:4240.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...