[Leu5]-Enkephalin
Supplier: Anaspec Inc
AS-24333
102996-548EA
134.46
USD
102996-548
[Leu5]-Enkephalin
Proteins and Peptides
Enkephalins are pentapeptides involved in regulating nociception in the body. There are two enkephalin forms, one containing leucine and the other containing methionine ("met"). Both are products of the proenkephalin gene.
Sequence:YGGFL
MW:555.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...