[Arg8]-Vasopressin (AVP) Peptide
Supplier: Anaspec Inc
AS-24289
AS-24290
102996-516EA
77.46
USD
102996-516
102996-518
[Arg8]-Vasopressin (AVP) Peptide
Proteins and Peptides
AVP have been identified as an important regulator of fluid and electrolyte homeostasis through its anti-diuretic action on the kidney, After myocardial infarction, plasma levels of [Arg8]-vasopressin rise to recover hemodynamics.
Sequence: CYFQNCPRG-NH2 (Disulfide bridge: 1-6)
MW: 1084.3 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...