Conus geographus Conantokin G
Supplier: Anaspec Inc
AS-24281
102996-508EA
454.54
USD
102996-508
Conus geographus Conantokin G
Proteins and Peptides
Conantokin G (Con G) toxin is a 17-amino-acid competitive antagonist of N-methyl-D-aspartate (NMDA) receptors. It was isolated from the venom of Conus geographus and it belongs to a unique family of g-carboxyglutamic acid-containing Conus peptides.
Sequence:GE(Gla)(Gla)LQ(Gla)NQ(Gla)LIR(Gla)KSN-NH2
MW:2264.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...