c-Myc peptide epitope
Supplier: Anaspec Inc
AS-24194
102996-474EA 61.38 USD
102996-474
c-Myc peptide epitope
Proteins and Peptides
c-Myc, the product of the c-myc proto-oncogene, is a helix-loop-helix leucine zipper phosphoprotein that regulates gene transcription in cell proliferation, cell differentiation and apoptosis. This peptide is a human c-myc epitope.
  • Sequence: EQKLISEEDL
References:
Johnson, G. and L. Hersh, J. Biol. Chem. 269, 7682 (1994); Chou, T. et al. J. Biol. Chem. 270, 18961 (1995); Kato, G. and C. Dang, FASEB J. 6, 3065 (1992). See also TNF-FRET 1 (#25349), TNF-FRET 2 (#27126).
Product Citations:
McLachlin, D. et al. (2003). Improved β-Elimination-Based Affinity Purification Strategy for Enrichment of Phosphopeptides. Anal Chem 75, 6826.