Anti-BetaGamma
Supplier: Anaspec Inc
AS-24177
AS-24178
102996-462EA
454.54
USD
102996-462
102996-464
Anti-BetaGamma
Proteins and Peptides
This is a membrane-permeable peptide, whose membrane-permeable sequence (MPS) is derived from the C-terminus of phosducin-like protein.
Sequence:AAVALLPAVLLALLAVTDQLGEDFFAVDLEAFLQEFGLLPEKE
MW:4601.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...