Kallikrein Substrate, AFC (Antibody-Fluorophore Conjugate)
Supplier: Anaspec Inc
AS-24137
102996-446EA
323
USD
102996-446
Kallikrein Substrate, AFC (Antibody-Fluorophore Conjugate)
Proteins and Peptides
This is a fluorescent glandular kallikrein substrate, Abs/Em=380/500 nm.
Sequence:vLR-AFC
MW:597.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...