Fluorogenic DPPIV Substrate, AFC (Antibody-fluorophore conjugate)
Supplier: Anaspec Inc
AS-24126
102996-438EA
279.15
USD
102996-438
Fluorogenic DPPIV Substrate, AFC (Antibody-fluorophore conjugate)
Proteins and Peptides
This is a fluorescent peptide, Abs/Em=380/500. It is a substrate for dipeptidyl peptidase IV (DPP IV) and Xaa-Pro dipeptidase.
Sequence:AP-AFC
MW:397.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...