Dipeptidylaminopeptidase IV Substrate, AMC (7-amino-4-methylcoumarin)
Supplier: Anaspec Inc
AS-24098
102996-430EA
239.69
USD
102996-430
Dipeptidylaminopeptidase IV Substrate, AMC (7-amino-4-methylcoumarin)
Proteins and Peptides
This is a fluorescent dipeptidylaminopeptidase IV substrate, Abs/Em=353/442 nm.
Sequence:GP-AMC
MW:329.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...