Human;Pig;Rat Viasoactive intestinal peptide, FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
AS-23614
102996-398EA
844.77
USD
102996-398
Human;Pig;Rat Viasoactive intestinal peptide, FAM (Carboxyfluorescein)
Proteins and Peptides
This is a fluorescent (FAM)-labeled VIP, Abs/Em=492/518 nm, a 28-amino acid neuropeptide which is a neurotransmitter and a neuromodulator.
Sequence:FAM-HSDAVFTDNYTRLRKQMAVKKYLNSILN-NH2
MW:3684.2 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...