Kemptide
Supplier: Anaspec Inc
AS-22594
AS-22595
102996-280EA
157.85
USD
102996-280
102996-282
Kemptide
Proteins and Peptides
Kemptide is a phosphate acceptor peptide that serves as a synthetic substrate for PKA (Km = 16 µM). The corresponding fluorescent and biotinylated peptides are also proven to be good substrates for PKA.
Sequence:LRRASLG
MW:771.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...