Human;Sheep;Rat PACAP (6-38), amide
Supplier: Anaspec Inc
AS-22516
AS-22517
102996-258EA
323
USD
102996-258
102996-260
Human;Sheep;Rat PACAP (6-38), amide
Proteins and Peptides
This peptide is a potent antagonist of PACAP 38 and is much more potent and selective than PACAP (6-27) in the inhibition of PACAP-27 stimulated pituitary adenylate cyclase. The Ki values for the inhibition of the enzyme are 7nM and 150 nM, respectively.
Sequence: FTDSYSRYRKQMAVKKYLAAVLGKRYKQRVKNK-NH2
MW: 4024.8 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...