Human;Rat Neuropeptide Y
Supplier: Anaspec Inc
AS-22464
AS-22465
102996-230EA
239.69
USD
102996-230
102996-232
Human;Rat Neuropeptide Y
Proteins and Peptides
Neuropeptide Y (NPY) is a 36aa neuropeptide involved in food intake, fat metabolism, stress and pain reduction, and vasodilation
Sequence:YPSKPDNPGEDAPAEDMARYYSALRHYINLITRQRY-NH2
MW:4271.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...