Toad Bombesin
Supplier: Anaspec Inc
AS-20665
102996-082EA
84.77
USD
102996-082
Toad Bombesin
Proteins and Peptides
Bombesin is a 14-aa peptide originally isolated from the fire-bellied toad. It has two mammalian homologs, neuromedin and Gastrin-Releasing Peptide (GRP). It binds with high affinity to the GRP receptor, a member of the G-protein-coupled receptor family. It stimulates gastrin release from G cells, and acts in the brain to stop eating behavior.
Bombesin is also a tumor marker for lung small cell carcinoma, neuroblastoma, pancreatic cancer and gastric cancer.
Sequence:Pyr-QRLGNQWAVGHLM-NH2
MW:1620.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...