Rat Atrial Natriuretic Peptide (1-28)
Supplier: Anaspec Inc
AS-20651
AS-20652
102996-078EA
254.31
USD
102996-078
102996-080
Rat Atrial Natriuretic Peptide (1-28)
Proteins and Peptides
Rat ANP differs from the human hormone by only one residue at position 12. ANP (1-28) peptide hormone constitutes the first 28 amino acids of the ANP synthesized in the heart of different vertebrates. It plays an important role in the regulation of blood pressure and natriuresis/diuresis. Residues Phe8, Arg14 and the C-terminal sequence of ANP are known to bind to human NPR-A (Natriuretic peptide receptor type A).
Sequence:SLRRSSCFGGRIDRIGAQSGLGCNSFRY (Disulfide bridge: 7-23)
MW:3062.5 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...