Human Atrial Natriuretic Peptide (1-28)

Supplier: Anaspec Inc

AS-20647 AS-20648
102996-074EA 254.31 USD
102996-074 102996-076
Human Atrial Natriuretic Peptide (1-28)
Proteins and Peptides

One of the three mammalian Natriuretic peptides, A-type is the Atrial natriuretic peptide (ANP) also called Cardiodilatin (CDD). ANP (1-28) peptide hormone constitutes the first 28 amino acids of the ANP synthesized in the heart of different vertebrates. It plays an important role in the regulation of blood pressure and natriuresis/diuresis. Residues Phe8, Arg14 and the C-terminal sequence of ANP are known to bind to human NPR-A (Natriuretic peptide receptor type A).
Sequence:SLRRSSCFGGRMDRIGAQSGLGCNSFRY (Disulfide bridge: 7-23)
MW:3080.5 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR