Chicken OVA (257-264)
Supplier: Anaspec Inc
AS-64413
102995-924EA
197.31
USD
102995-924
Chicken OVA (257-264)
Proteins and Peptides
FILKSINE is the scrambled version of ovalbumin (OVA) peptide (257-264), SIINFEKL. OVA Peptide is a class I (Kb)-restricted peptide epitope of ovalbumin presented by the class I MHC (major histocompatibility complex) molecule, H-2Kb (class I genes of the mouse MHC).
Sequence: FILKSINE
MW: 963.2 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...