ER 172594-06 hydrobromide ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Atopaxar hydrobromide

SYN-3002-M001 SYN-3002-M005 SYN-3002-M010
102992-840EA 292.15 USD
102992-840 102992-842 102992-844
ER 172594-06 hydrobromide ≥95%
ER 172594-06 hydrobromide

E5555 shows potent inhibitory effects on human platelet aggregation induced by thrombin and TRAP with IC values of 0.064 and 0.031µM, respectively.

Formula: C₂₉H₃₉BrFN₃O₅·HBr
Storage Temperature: Freezer
CAS Number: 474550-69-1

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR