GDC-0032 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: Taselisib, 2-(4-(2-(1-Isopropyl-3-methyl-1H-1,2,4-triazol-5-yl)-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepin-9-yl)-1H-pyrazol-1-yl)-2-methylpropanamide

SYN-1202-M001 SYN-1202-M005 SYN-1202-M010 SYN-1202-M050
102992-762EA 119.08 USD
102992-762 102992-764 102992-766 102992-768
GDC-0032 ≥95%
GDC-0032

GDC-0032 is a potent, next-generation beta isoform-sparing PI3K inhibitor targeting PI3Kalpha/delta/gamma with Ki of 0.29nM/0.12nM/0.97nM, 10 fold selective over PI3Kbeta.

Formula: C₂₄H₂₈N₈O₂
Storage Temperature: Freezer
MDL Number: MFCD28167893
CAS Number: 1282512-48-4

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Slightly soluble (1mg/ml) in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR