PF-04979064 ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1194-M001 SYN-1194-M005 SYN-1194-M010
102992-702EA 292.15 USD
102992-702 102992-704 102992-706
PF-04979064 ≥95%
PF-04979064

PF-04979064 is a potent and selective PI3K/mTOR dual kinase inhibitor with an IC(50) of 9.1nM for the PI3K family when tested AKT phosphorylation in BT20 cells. In direct kinase assays, PF-04979064 has potent Ki values against PI3Kalpha (Ki= 0.13nM human, 0.299nM mouse) and PI3Kgamma (Ki=0.111nM), and PI3Kdelta (Ki=0.122nM). In contrast, Ki values for non-PI3K kinases such as mTOR were 10X higher at 1.42 nM.

Formula: C₂₄H₂₆N₆O₃
Storage Temperature: Refrigerator
CAS Number: 1220699-06-8

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR