MPI-0479605 hydrochloride ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1187-M001 SYN-1187-M005 SYN-1187-M010
102992-654EA 283.85 USD
102992-654 102992-656 102992-658
MPI-0479605 hydrochloride ≥95%
MPI-0479605 hydrochloride

MPI-0479605 inhibits TTK kinase activity in vitro with an IC(50) of 4nM.

Formula: C₂₂H₂₉N₇O·HCl
Storage Temperature: Refrigerator
CAS Number: 1246529-32-7

Order Now

Specification Test Results

Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR