CX-6258 ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

SYN-1182-M001 SYN-1182-M005 SYN-1182-M010
102992-620EA 283.85 USD
102992-620 102992-622 102992-624
CX-6258 ≥98% (by HPLC)
CX-6258

CX6258 is a potent inhibitor of Pim-1, Pim-2 and Pim-3 with IC(50) values of 5, 25 and 16nM, respectively.

Formula: C₂₆H₂₄ClN₃O₃
Storage Temperature: Freezer
CAS Number: 1202916-90-2

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR