AZD-8055 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (5-(2,4-Bis((S)-3-methylmorpholino)pyrido[2,3-d]pyrimidin-7-yl)-2-methoxyphenyl)methanol, AZD 8055, (5-(2,4-bis((3s)-3-methylmorpholin-4-yl)pyrido(2,3-d)pyrimidin-7-yl)-2-methoxyphenyl)methanol, AZD8055, CCG168

SYN-1166-M001 SYN-1166-M005 SYN-1166-M010 SYN-1166-M050
102992-496EA 90 USD
102992-496 102992-498 102992-500 102992-502
AZD-8055 ≥95%
AZD-8055

AZD8055 is a novel ATP-competitive inhibitor of mTOR kinase activity with an IC(50) of 0.8nM. It shows excellent selectivity (~1,000-fold) against all class I phosphatidylinositol 3-kinase (PI3K) isoforms and other members of the PI3K-like kinase family.

Formula: C₂₅H₃₁N₅O₄
Storage Temperature: Freezer
MDL Number: MFCD16660191
CAS Number: 1009298-09-2

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR