SAR-131675 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: (-)-2-Amino-1-ethyl-7-(3-hydroxy-4-methoxy-3-methylbut-1-yn-1-yl)-N-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxamide

SYN-1165-M001 SYN-1165-M005 SYN-1165-M010
102992-490EA 283.85 USD
102992-490 102992-492 102992-494
SAR-131675 ≥95%
SAR-131675

SAR131675 is a potent and selective VEGFR-3 inhibitor. It inhibited VEGFR-3 tyrosine kinase activity and VEGFR-3 autophosphorylation in HEK cells with IC(50) values of 20 and 45nM, respectively. SAR131675 dose dependently inhibited the proliferation of primary human lymphatic cells, induced by the VEGFR-3 ligands VEGFC and VEGFD, with an IC(50) of about 20nM. SAR131675 was found to be highly selective for VEGFR-3 versus 107 receptors, enzymes, ion channels, and 65 kinases. However, it was moderately active on VEGFR-2 with a VEGFR-3/VEGFR-2 ratio of about 10. SAR131675 had no antiproliferative activity on a panel of 30 tumors and primary cells, further showing its high specificity and indicating that SAR131675 is not a cytotoxic or cytostatic agent.

Formula: C₁₈H₂₂N₄O₄
Storage Temperature: Freezer
MDL Number: MFCD23098773
CAS Number: 1092539-44-0

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR