PF-4618433 ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1163-M001 SYN-1163-M005 SYN-1163-M010
102992-478EA 283.85 USD
102992-478 102992-480 102992-482
PF-4618433 ≥95%
PF-4618433

Potent and selective PYK2 inhibitor. PF-4618433 shows improved PYK2 potency, reduced p38 activity, and superior overall selectivity relative to the prototype BIRB796. In the functional osteogenesis assays, cultures treated with increasing concentrations of PF-4618433 starting at day 1 and maintained throughout showed a dose-dependent increases in both alkaline phosphatase activity and mineralization.

Formula: C₂₄H₂₇N₇O₂
Storage Temperature: Freezer
CAS Number: 1166393-85-6

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR