NVP-BGJ398 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 3-(2,6-Dichloro-3,5-dimethoxyphenyl)-1-[6-[4-(4-ethylpiperazin-1-yl)anilino]pyrimidin-4-yl]-1-methylurea, 3-(2,6-Dichloro-3,5-dimethoxyphenyl)-1-(6-((4-(4-ethylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)-1-methylurea, Infigratinib

SYN-1152-M001 SYN-1152-M005 SYN-1152-M010 SYN-1152-M050
102992-398EA 90 USD
102992-398 102992-400 102992-402 102992-404
NVP-BGJ398 ≥95%
NVP-BGJ398

NVP-BGJ398 inhibits FGFR-WT, FGFR2-WT, FGFR3-K650E, FGFR3-S249C and FGFR4-WT with IC(50) values of 4.6, 4.9, 5, 5 and 168nM, respectively.

Formula: C₂₆H₃₁Cl₂N₇O₃
Storage Temperature: Freezer
MDL Number: MFCD22123241
CAS Number: 872511-34-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR