Cabozantinib ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide, XL184

SYN-1138-M001 SYN-1138-M005 SYN-1138-M010 SYN-1138-M050
102992-304EA 96.92 USD
102992-304 102992-306 102992-308 102992-310
Cabozantinib ≥95%
Cabozantinib

Cabozantinib is very potent inhibitor, with an IC(50) of less than 10nM for VEGFR2, MET, Kit and FLT3-ITD.

Formula: C₂₈H₂₄FN₃O₅
Storage Temperature: Freezer
MDL Number: MFCD20926324
CAS Number: 849217-68-1

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR